Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Fentanyl Drugs of Abuse Assay
GSA_VA

Catalog No. 23278369 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3 x 18 mL
1 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-369 3 x 18 mL
21-200-421 1 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Criminal Justice and Forensic use only.
Catalog No. 23-278-369 Supplier Thermo Scientific™ Supplier No. 10016006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Criminal Justice and Forensic use only.

Save time while maintaining accuracy with these liquid, ready-to-use assays, which are for Criminal Justice and Forensic use only and are for sale in the U.S.A. only.

Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Fentanyl Drugs of Abuse Assays. Field-tested on a wide range of clinical analyzers, including the Thermo Scientific™ Indiko™ Clinical Analyzer, these liquid, ready-to-use assays have been widely accepted in laboratories where cost control and optimal productivity are key issues. These assays are for Criminal Justice and Forensic use only and are for sale in the U.S.A. only.

  • Specific detection of fentanyl
  • Qualitative or semi-quantitative modes  
  • Excellent correlation to LC-MS/MS
  • Applications for an array of clinical chemistry analyzers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets DRI Fentanyl Control Sets
Description DRI Fentanyl Drugs of Abuse Assay for Indiko. Criminal Justice and Forensic use only. For sale in the U.S.A. only.
Detectable Analytes Fentanyl (FEN)
DoA Calibrators Fentanyl Calibrator
Sample Type Urine
Type Reagent
Quantity 3 x 18 mL

For sale in the U.S.A. only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.