Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Assay

Catalog No. 10028872 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 500 mL 
1 x 115 mL 
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-028-872 1 x 115 mL 
10-028-871 1 x 500 mL 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-028-872 Supplier Thermo Scientific™ Supplier No. 10028872
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifically detect fentanyl and its primary metabolite, norfentanyl in urine using the Thermo Scientific™ DRI™ Fentanyl II drugs of abuse assay. This automated fentanyl urine drug screening test delivers quick and accurate preliminary results, enabling timely interventions for fentanyl abuse.

Developed for hospital laboratories, addiction treatment centers, and toxicology laboratories, the Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Assay is an effective tool for the screening fentanyl abuse and misuse. Its ability to detect both fentanyl and its primary metabolite, norfentanyl gives this assay a broader window of detection compared to competing products. This automated urine drug screening assay exhibits negligible cross-reactivity to other opioids. It is the only 510(k) cleared assay that combines norfentanyl cross-reactivity with a low 1 ng/mL cutoff concentration.

  • Specifically detect fentanyl and its major metabolite, norfentanyl, at physiologically relevant concentrations in human urine 
  • Low cross-reactivity to other non-fentanyl synthetic opioids and over-the-counter medication 
  • High sensitivity with a cutoff concentration of 1 ng/mL 
  • Superior assay precision 
  • Excellent correlation to LC-MS/MS 
  • Liquid, Ready-to-Use 
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets DRI Fentanyl II Control Kit 
Description DRI Fentanyl II Calibrators 
Detectable Analytes Fentanyl (FEN)
DoA Calibrators Fentanyl II Calibrator
Quantity 1 x 115 mL 

For sale in the U.S.A. and Canada only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.