Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Calibrators

Catalog No. 10028873
Change view
Click to view available options
Description:
DRI Fentanyl II Cutoff Calibrator 
DRI Fentanyl II Negative Calibrator
Concentration:
0 ng/mL 
1 ng/mL 
Detectable Analytes:
Fentanyl
Negative Calibrator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Concentration Detectable Analytes
10-028-873 DRI Fentanyl II Cutoff Calibrator  1 ng/mL  Fentanyl
10-028-874 DRI Fentanyl II Negative Calibrator 0 ng/mL  Negative Calibrator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-028-873 Supplier Thermo Scientific™ Supplier No. 10028873
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ensure accurate calibration with the Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Calibrators, designed for urine screening with the Thermo Scientific DRI Fentanyl II Drugs of Abuse Assay.

Ensure accurate calibration with the Thermo Scientific DRI Fentanyl II Drugs of Abuse Calibrators, designed for urine screening with the Thermo Scientific DRI Fentanyl II Drug of Abuse Assay. The calibrators, sold separately, include a negative urine calibrator and a cutoff calibrator spiked with 1 ng/mL of fentanyl.

  • Specifically detect fentanyl and its major metabolite, norfentanyl, at physiologically relevant concentrations in human urine
  • Low cross-reactivity to other non-fentanyl synthetic opioids and over-the-counter medication 
  • High sensitivity with a cutoff concentration of 1 ng/mL 
  • Superior assay precision
  • Excellent correlation to LC-MS/MS
  • Liquid, Ready-to-Use 
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI Fentanyl II Cutoff Calibrator 
Concentration 1 ng/mL 
Detectable Analytes Fentanyl
Sample Type Urine
Packaging 2 Bottles
Quantity 2 x 10 mL 

For sale in the U.S.A. and Canada only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.