Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Control Set

Catalog No. 10028875 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 Vials/Level, 10 mL Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-028-875 2 Vials/Level, 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-028-875 Supplier Thermo Scientific™ Supplier No. 10028875
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensure accurate calibration with the Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Control Set, designed for urine screening with the Thermo Scientific DRI Fentanyl II Drugs of Abuse Assay.

Ensure accurate calibration with the Thermo Scientific DRI Fentanyl II High and Low Control Set, designed for urine screening with the Thermo Scientific DRI Fentanyl II Drug of Abuse Assay. The control set includes a high and low control, targeted at 1.5 ng/mL and 0.5 ng/mL respectively, to validate the calibration curve by providing positive and negative results.

  • Specifically detect fentanyl and its major metabolite, norfentanyl, at physiologically relevant concentrations in human urine
  • Low cross-reactivity to other non-fentanyl synthetic opioids and over-the-counter medication 
  • High sensitivity with a cutoff concentration of 1 ng/mL 
  • Superior assay precision
  • Excellent correlation to LC-MS/MS
  • Liquid, Ready-to-Use 
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI Fentanyl II Control Kit 
Concentration 0.5 ng/mL Low, 1.5 ng/mL High
Detectable Analytes Fentanyl
Sample Type Urine
Type Control
Level High, Low
Packaging 4 Bottles
Quantity 2 Vials/Level, 10 mL Each

For sale in the U.S.A. and Canada only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.