Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ General Oxidant-Detect™ Specimen Validity Test Calibrators

Catalog No. 23278342 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-342 2 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-342 Supplier Thermo Scientific™ Supplier No. 10009971
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensure accuracy in testing for specimen adulterants with the liquid, ready-to-use Thermo Scientific™ DRI™ General Oxidant-Detect™ Specimen Validity Test Calibrators.

Ensure accuracy in testing for specimen adulterants with the liquid, ready-to-use Thermo Scientific™ DRI™ General Oxidant-Detect™ Specimen Validity Test Calibrators, for use on a variety of general chemistry analyzers.

The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution. All of our urine adulteration assays are liquid, ready-to-use and can be performed on a variety of general chemistry analyzers. The adulteration assays provide a complete and automated way to test for sample integrity and adulteration.

Features:
  • Most extensive menu of tests available
  • Offered in convenient sizes
  • Liquid, ready-to-use
  • Can be used on a variety of general chemistry analyzers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI™ General Oxidant Specimen Validity Test Calibrator Set
Concentration 0, 200 μg/mL
Detectable Analytes General Oxidants
Sample Type Urine
Type Calibrator
Quantity 2 x 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.