Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ General Oxidant-Detect™ Specimen Validity Test

Catalog No. 23278341 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-341 2 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-341 Supplier Thermo Scientific™ Supplier No. 10009958
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform liquid, ready-to-use DRI General Oxidant-Detect Specimen Validity Tests on a variety of general chemistry analyzers.

Perform liquid, ready-to-use Thermo Scientific™ DRI™ General Oxidant-Detect™ Specimen Validity Tests on a variety of general chemistry analyzers.

Our adulteration assays provide a complete and automated way to test for sample integrity and adulteration.
  • One of the most extensive menus of tests available
  • Associated calibrators and controls offered in convenient sizes
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI General Oxidant-Detect Assay Specimen Validity Test
Detectable Analytes General Oxidants
Sample Type Urine
Type Reagent
Quantity 2 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.