Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators

Catalog No. 23278423 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
1.010
1.025
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-278-423 1.010
23-278-424 1.025
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-278-423 Supplier Thermo Scientific™ Supplier No. 1754
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators.

Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators. Liquid and ready-to-use, these controls can be used with the Thermo Scientific™ DRI™ Gravity-Detect™ Reagent Kit on a variety of general chemistry analyzers. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution.

  • Can be performed on a variety of general chemistry analyzers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Description DRI™ Gravity-Detect™ Specimen Validity Test Calibrators
Detectable Analytes Specific Gravity
Sample Type Urine
Type Calibrator
Level 1.010
Quantity 1 x 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.