Learn More
Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators
Catalog No. 23278424 Shop All Thermo Scientific Products
Change view
Click to view available options
Level:
1.010
1.025
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-278-424 1.025
23-278-423 1.010
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-278-424 Supplier Thermo Scientific™ Supplier No. 1755
Add to Cart
Add to Cart
This item is not returnable. View return policy
Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators.
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Calibrators. Liquid and ready-to-use, these controls can be used with the Thermo Scientific™ DRI™ Gravity-Detect™ Reagent Kit on a variety of general chemistry analyzers. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution.
  • Can be performed on a variety of general chemistry analyzers
TRUSTED_SUSTAINABILITY
Specifications
Description DRI™ Gravity-Detect™ Specimen Validity Test Calibrators
Detectable Analytes Specific Gravity
Sample Type Urine
Type Calibrator
Level 1.025
Quantity 1 x 25 mL