Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Controls

Catalog No. 23278425 Shop All Thermo Scientific Products
Change view
Click to view available options
Concentration:
1.015 μg/mL
1.030 μg/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
23-278-425 1.015 μg/mL
23-278-426 1.030 μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-278-425 Supplier Thermo Scientific™ Supplier No. 1756
Add to Cart
Add to Cart
This item is not returnable. View return policy

Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Controls.

Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Controls. Liquid and ready-to-use, these controls can be performed on a variety of general chemistry analyzers. The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution.

Our adulteration assays provide a complete and automated way to test for sample integrity and adulteration.
  • Associated calibrators and controls offered in convenient sizes
  • Liquid, ready-to-use
  • Can be performed on a variety of general chemistry analyzers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Description Gravity-Detect™
Concentration 1.015 μg/mL
Detectable Analytes Specific Gravity
Sample Type Urine
Type Control
Quantity 1 x 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.