Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Control Set

Catalog No. 23278381 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-381 2 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-381 Supplier Thermo Scientific™ Supplier No. 10018149
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Save time while maintaining accuracy with the liquid, ready-to-use Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Control Set.

Streamline your testing process while maintaining high analytical accuracy by utilizing the Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Control Set along with our Thermo Scientific DRI Hydrocodone Drugs of Abuse Assay and Calibrators. This liquid, ready-to-use control set is for sale in the U.S.A. and Canada only.

  • Specific detection of hydrocodone
  • Qualitative or semi-quantitative modes for results
  • For sale in the U.S.A. and Canada only
TRUSTED_SUSTAINABILITY

Specifications

Description DRI Hydrocodone Control Set
Concentration 300 ng/mL cutoff, Low (225ng/mL), High (375ng/mL)
Detectable Analytes Hydrocodone
Sample Type Urine
Type Control
Level High, Low
Quantity 2 x 10 mL

Disclaimers: Some of these products are not cleared in every country. Please contact your sales representative for more information. Product claims may differ from country to country based on regulations and product approvals. Please contact your country representative for further details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.