Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Methadone Drugs of Abuse Assays

Catalog No. 23278375 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3 x 18 mL
1 x 100 mL
1 x 500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-375 3 x 18 mL
23-278-325 1 x 100 mL
21-200-416 1 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-278-375 Supplier Thermo Scientific™ Supplier No. 10016403
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use Thermo Scientific™ DRI™ Methadone Drugs of Abuse Assays to achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation.

Use Thermo Scientific™ DRI™ Methadone Drugs of Abuse Assays to achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation. These liquid, ready-to-use assays offer applications for an array of clinical chemistry analyzers. Field-tested on a wide range of analyzers, these assays are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets MGC Multi-Drug Controls
Description DRI Methadone Drugs of Abuse Assay for Indiko
Detectable Analytes Methadone (MTD)
DoA Calibrators Multi-drug Calibrator
Sample Type Urine
Type Reagent
Quantity 3 x 18 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.