Learn More
Description
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation when you choose Thermo Scientific™ DRI™ Opiate Drugs of Abuse Calibrators. These ready-to-use assays in liquid form have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Applications for an array of clinical chemistry analyzers
- Variety of calibrators and multiconstituent control sets
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | DRI; Opiate Calibrator 3 |
| Concentration | 500 ng/mL |
| Detectable Analytes | Opiate |
| Sample Type | Urine |
| Type | Calibrator |
| Packaging | 1 Bottle |
| Quantity | 1 x 25 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.