Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Assays
GSA_VA

Catalog No. 23278385 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 68 mL
3 x 18 mL
1 x 500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-385 1 x 68 mL
21-200-432 1 x 500 mL
23-278-355 3 x 18 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-278-385 Supplier Thermo Scientific™ Supplier No. 100248
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Assays.

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs. Supports new SAMHSA guidelines (effective October 1, 2017)

  • One of the largest menu of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets DRI Oxycodone Control Set
Description DRI Oxycodone Drugs of Abuse Assays
Detectable Analytes Oxycodone (OXY)
DoA Calibrators Oxycodone Calibrator
Sample Type Urine
Type Reagent
Quantity 1 x 68 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.