Learn More
Description
Eliminate time-consuming steps while achieving high analytical accuracy when you choose the ready-to-use Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Control Set in liquid form. These assays have been field-tested on a wide range of analyzers, and they are enthusiastically accepted in laboratories where cost-control and optimal productivity are key issues.
- One of the largest menus of tests available
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Applications for an array of clinical chemistry analyzers
- Variety of calibrators and multiconstituent control sets
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | DRI Oxycodone Drugs of Abuse Control Set |
| Concentration | 100 ng/mL |
| Detectable Analytes | Oxycodone |
| Sample Type | Urine |
| Type | Control |
| Level | High (125ng/mL) and Low (75ng/mL) |
| Quantity | 2 x 10 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
