Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ pH-Detect™ Specimen Validity Tests
GSA_VA

Catalog No. 23278335 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-335 2 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-335 Supplier Thermo Scientific™ Supplier No. 100054
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ pH-Detect Specimen Validity Tests.

Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ pH-Detect Specimen Validity Tests. Liquid and ready-to-use, these tests can be performed on a variety of general chemistry analyzers.

The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution. The adulteration assays provide a complete and automated way to test for sample integrity and adulteration.

  • Complete and automated way to test for sample integrity and adulteration
  • Associated calibrators and controls offered in convenient sizes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI pH-Detect Assay Specimen Validity Test
Detectable Analytes pH
Sample Type Urine
Type Reagent
Quantity 2 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.