Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Propoxyphene Drugs of Abuse Assay

Catalog No. 23278321
Change view
Click to view available options
Quantity:
3 x 18 mL
1 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-321 1 x 100 mL
23-278-382 3 x 18 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-278-321 Supplier Thermo Scientific™ Supplier No. 0432
Add to Cart
Add to Cart
This item is not returnable. View return policy

Eliminate time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Propoxyphene Drugs of Abuse Assay.

Eliminate time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Propoxyphene Drugs of Abuse Assay. This liquid, ready-to-use assay delivers high analytical accuracy for an array of clinical chemistry analyzers. This liquid, ready-to-use assay has been field-tested on a wide range of analyzers. It is enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets MGC Primary DAU Control Set
Description DRI Propoxyphene Drugs of Abuse Assay
Detectable Analytes Propoxyphene (PPX)
DoA Calibrators Multi-drug Calibrator
Sample Type Urine
Type Reagent
Quantity 1 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.