Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI Tramadol Calibrator Set

Catalog No. 21200446 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 10 mL
5 x 10 mL
Concentration:
0 ng/mL
0, 100, 200, 500 and 1000 ng/mL
100 ng/mL
Type:
Calibrator Set
Cutoff Calibrator
Negative Calibrator
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration Type
21-200-446 5 x 10 mL 0, 100, 200, 500 and 1000 ng/mL Calibrator Set
21-200-443 2 x 10 mL 100 ng/mL Cutoff Calibrator
21-200-444 2 x 10 mL 0 ng/mL Negative Calibrator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 21-200-446 Supplier Thermo Scientific™ Supplier No. B40000366
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ensure accurate calibration with the Thermo Scientific DRI Tramadol High and Low Control Set, designed for urine screening with the Thermo Scientfic DRI Tramadol Drugs of Abuse Assay. The control set includes a high and low control targeted at 75 ng/mL and 125 ng/mL respectively, to validate the calibration curve by providing positive and negative results.

  • Specifically detect tramadol and its primary metabolites O-Desmethyltramadol and N-Desmethyltramadol at physiologically relevant concentrations in human urine
  • Low cross-reactivity to non-tramadol compounds
  • High sensitivity with a cutoff concentration of 100 ng/mL
  • Superior assay precision
  • Excellent correlation to LC-MS/MS
  • Liquid, Ready-to Use
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance FDA Cleared
Description Semi-quantitative
Concentration 0, 100, 200, 500 and 1000 ng/mL
Detectable Analytes Tramadol
For Use With (Equipment) Automated Chemistry Analyzers
Sample Type Urine
Type Calibrator Set
Quantity 5 x 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.