Learn More
Description
Ensure accurate calibration with the Thermo Scientific DRI Tramadol High and Low Control Set, designed for urine screening with the Thermo Scientfic DRI Tramadol Drugs of Abuse Assay. The control set includes a high and low control targeted at 75 ng/mL and 125 ng/mL respectively, to validate the calibration curve by providing positive and negative results.
- Specifically detect tramadol and its primary metabolites O-Desmethyltramadol and N-Desmethyltramadol at physiologically relevant concentrations in human urine
- Low cross-reactivity to non-tramadol compounds
- High sensitivity with a cutoff concentration of 100 ng/mL
- Superior assay precision
- Excellent correlation to LC-MS/MS
- Liquid, Ready-to Use
Specifications
Specifications
| Description | DRI Tramadol Drugs of Abuse Control Set |
| Concentration | 75 ng/mL Low, 125 ng/mL High |
| Detectable Analytes | Tramadol |
| For Use With (Equipment) | Automated Chemistry Analyzers |
| Sample Type | Urine |
| Type | Control Set |
| Intended Use | Quality control of the DRI Tramadol Assay |
| Packaging | 4 bottles |
| Quantity | 2 Vials/Levels, 10 mL Each |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.