Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI Tramadol Drugs of Abuse Assay

Catalog No. 21200441
Change view
Click to view available options
Quantity:
1 x 115mL
1 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-200-441 1 x 500 mL
21-200-442 1 x 115mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-200-441 Supplier Thermo Scientific™ Supplier No. B40000350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Developed for hospital laboratories, addiction treatment centers, and toxicology laboratories, the Thermo Scientific™ DRI™ Tramadol Drugs of Abuse Assay is an effective tool for the screening of tramadol abuse and misuse.

Developed for hospital laboratories, addiction treatment centers, and toxicology laboratories, the Thermo Scientific™ DRI™ Tramadol Drugs of Abuse Assay is an effective tool for the screening tramadol abuse and misuse. Its ability to detect tramadol and its primary metabolites, O-Desmethyltramadol and N-Desmethyltramadol gives this assay a broader window of detection compared to competing products. This automated urine drug screening assay exhibits negligible cross-reactivity to other structurally related compounds. It is the only on-market 510(k) cleared assay that demonstrates a cross-reactivity with a low 100 ng/mL cutoff concentration.

  • Specifically detect tramadol and its primary metabolites O-Desmethyltramadol and N-Desmethyltramadol at physiologically relevant concentrations in human urine
  • Low cross-reactivity to non-tramadol compounds
  • High sensitivity with a cutoff concentration of 100 ng/mL
  • Superior assay precision • Excellent correlation to LC-MS/MS
  • Liquid, Ready-to Use
TRUSTED_SUSTAINABILITY

Specifications

Control Sets DRI Tramadol Drugs of Abuse Control Kit
Description DRI Tramadol Drugs of Abuse Assay
Detectable Analytes Tramadol (TRA)
DoA Calibrators No
Format Kit
Sample Type Urine
Type Urine
For Use With (Application) Enzyme Immunoassay Detection
Quantity 1 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.