Learn More
Description
Developed for hospital laboratories, addiction treatment centers, and toxicology laboratories, the Thermo Scientific™ DRI™ Tramadol Drugs of Abuse Assay is an effective tool for the screening of tramadol abuse and misuse.
Developed for hospital laboratories, addiction treatment centers, and toxicology laboratories, the Thermo Scientific™ DRI™ Tramadol Drugs of Abuse Assay is an effective tool for the screening tramadol abuse and misuse. Its ability to detect tramadol and its primary metabolites, O-Desmethyltramadol and N-Desmethyltramadol gives this assay a broader window of detection compared to competing products. This automated urine drug screening assay exhibits negligible cross-reactivity to other structurally related compounds. It is the only on-market 510(k) cleared assay that demonstrates a cross-reactivity with a low 100 ng/mL cutoff concentration.
- Specifically detect tramadol and its primary metabolites O-Desmethyltramadol and N-Desmethyltramadol at physiologically relevant concentrations in human urine
- Low cross-reactivity to non-tramadol compounds
- High sensitivity with a cutoff concentration of 100 ng/mL
- Superior assay precision • Excellent correlation to LC-MS/MS
- Liquid, Ready-to Use
Specifications
Specifications
| Control Sets | DRI Tramadol Drugs of Abuse Control Kit |
| Description | DRI Tramadol Drugs of Abuse Assay |
| Detectable Analytes | Tramadol (TRA) |
| DoA Calibrators | No |
| Format | Kit |
| Sample Type | Urine |
| Type | Reagent |
| For Use With (Application) | Enzyme Immunoassay Detection |
| Quantity | 1 x 115mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.