Learn More
Thermo Scientific™ DRI Tricyclics Serum Toxicology Assays
Catalog No. 09980609 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 X 500 mL
25 x 8 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-980-609 1 X 500 mL
23-278-399 25 x 8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-980-609 Supplier Thermo Scientific™ Supplier No. 10028827
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quickly and accurately identify tricyclics in serum samples with Thermo Scientific™ DRI™ Tricyclics Serum Toxicology Assays, excellent tools for emergency rooms when urine specimens are not conveniently available.
These Serum Toxicology Tricyclics Assays have the flexibility of analyzing serum or urine samples. These assays are provided in convenient packaging with separate calibrators for ease of use. A multi-analyte Serum Tox Control is also available. The assays are liquid, ready-to-use and have the versatility to be run on a variety of clinical chemistry analyzers.
  • Serum, plasma-and urine-based assay
  • Used to identify class of drug overdose
TRUSTED_SUSTAINABILITY
Specifications
Description DRI Tricyclics Serum Toxicology Assays
Detectable Analytes Tricyclic Antidepressants (TCA)
Sample Type Serum, Plasma, Urine
Type Reagent
Quantity 1 X 500 mL
For sale in the U.S.A. and Canada only.