Learn More
Description
Quickly and accurately identify tricyclics in serum samples with Thermo Scientific™ DRI™ Tricyclics Serum Toxicology Assays, excellent tools for emergency rooms when urine specimens are not conveniently available.
- Serum-and urine-based assay
- Used to identify class of drug overdose
Ordering Alerts:
1128: Reagent volume 25, 8mL.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | DRI Tricyclics |
| Detectable Analytes | Tricyclic Antidepressants (TCA) |
| Sample Type | Serum |
| Type | Reagent |
| Quantity | 25 x 8 mL |
For sale in the U.S.A. and Canada only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.