Learn More
AdipoGen DRibose
SDP
Catalog No. 502560115
Encompass_Preferred
Chemical. CAS 50-69-1. Formula C5H10O5. MW 150.13. Synthetic. Aldopentose monosaccharide that is phosphorylated into D-ribose 5-phosphate by ribokinase. D-ribose-5-phosphate supports the biosynthesis of the amino acids tryptophan and histidine, and is a component in the pentose phosphate pathway. D-ribose is present in all living cells and their microenvironments and is a key component of numerous biomolecules involved in many important metabolic pathways. It also participates in the glycation of proteins producing advanced glycation end products AGEs that lead to cell dysfunction and death. Ribosylation, has been shown to cause protein aggregation in vitro and in vivo. Has demonstrated significant enhancing abilities in replenishing deficient cellular energy levels following myocardial ischemia, improving depressed function in numerous animal investigations and showing cellular protection during oxidative stress.
Catalog No. 50-256-0115 Supplier AdipoGen Supplier No. CDXR0065G100
Chemical. CAS 50-69-1. Formula C5H10O5. MW 150.13. Synthetic. Aldopentose monosaccharide that is phosphorylated into D-ribose 5-phosphate by ribokinase. D-ribose-5-phosphate supports the biosynthesis of the amino acids tryptophan and histidine, and is a component in the pentose phosphate pathway. D-ribose is present in all living cells and their microenvironments and is a key component of numerous biomolecules involved in many important metabolic pathways. It also participates in the glycation of proteins producing advanced glycation end products AGEs that lead to cell dysfunction and death. Ribosylation, has been shown to cause protein aggregation in vitro and in vivo. Has demonstrated significant enhancing abilities in replenishing deficient cellular energy levels following myocardial ischemia, improving depressed function in numerous animal investigations and showing cellular protection during oxidative stress.