Learn More
Medchemexpress LLC Dried aluminum hydro 10mg
SDP
Catalog No. 5000206481
Encompass_Preferred
Dried aluminum hydroxide gel is a commonly used antacid with significant compound adhesion and release activity Dried aluminum hydroxide gel can effectively neutralize gastric acid and relieve stomach discomfort Dried aluminum hydroxide gel is also often used as an adjuvant in vaccines to enhance immune response Dried aluminum hydroxide gel can improve the stability and bioavailability of compounds in preparations
Catalog No. 50-002-06481 Supplier Medchemexpress LLC Supplier No. HYW116335A10MG
Dried aluminum hydroxide gel is a commonly used antacid with significant compound adhesion and release activity Dried aluminum hydroxide gel can effectively neutralize gastric acid and relieve stomach discomfort Dried aluminum hydroxide gel is also often used as an adjuvant in vaccines to enhance immune response Dried aluminum hydroxide gel can improve the stability and bioavailability of compounds in preparations