Learn More
Bioworld Dried Blood Spot (DBS) Genomic DNA Isolation Kit, 50 Prep(s)
Catalog No. 50196944
Encompass_Preferred
Rapid preparation of total genomic DNA from dried blood spots High quality DNA, free from RNA contamination Isolate genomic DNA from anticoagulated and untreated blood Rapid and convenient spin column procedure FEATURES AND BENEFITSFast and easy processing - Rapid spin-column format allows for the isolation of DNA from dried blood spots in 30 minutes High quality DNA - Total genomic DNA is of the highest quality and free from RNA contamination Isolate genomic DNA from a diversity of species - Genomic DNA can be isolated from the blood of various species, including humans Recovered genomic DNA is suitable for downstream applications - Purified genomic DNA is fully compatible with PCR amplifications and Southern Blot analysis
Catalog No. 50-196-944 Supplier Bioworld Supplier No. 107600871
Rapid preparation of total genomic DNA from dried blood spots High quality DNA, free from RNA contamination Isolate genomic DNA from anticoagulated and untreated blood Rapid and convenient spin column procedure FEATURES AND BENEFITSFast and easy processing - Rapid spin-column format allows for the isolation of DNA from dried blood spots in 30 minutes High quality DNA - Total genomic DNA is of the highest quality and free from RNA contamination Isolate genomic DNA from a diversity of species - Genomic DNA can be isolated from the blood of various species, including humans Recovered genomic DNA is suitable for downstream applications - Purified genomic DNA is fully compatible with PCR amplifications and Southern Blot analysis