Learn More
WA HAMMOND DRIERITE COMPANY DRIERITE, REGULAR (NON-INDICATING) 8 MESH 1 LB JAR, STOCK# 13001
Catalog No. NC3372145
Encompass_Preferred
Regular Drierite is anhydrous calcium sulfate. It is of neutral pH, constant in volume, chemically inert except toward water, insoluble in organic liquids and refrigerants, non-disintegrating, non-wetting, non-toxic, non-corrosive, non-channeling, generally recognized as safe (GRAS) by the FDA, not regulated by OSHA, economical to use, and can be regenerated repeatedly for reuse. Efficiency: The moisture remaining in gases dried with Drierite at 25° - 30° C is 0.005 mg./liter, (data verified by the National Bureau of Standards). Air is dried to a dew point of -100° F. Organic liquids are dried to the low ppm range. There is little change in efficiency at temperatures up to 100° C. Typical Analysis: >99.5% CaSO4
Catalog No. NC3372145 Supplier WA HAMMOND DRIERITE COMPANY Supplier No. 13001
Regular Drierite is anhydrous calcium sulfate. It is of neutral pH, constant in volume, chemically inert except toward water, insoluble in organic liquids and refrigerants, non-disintegrating, non-wetting, non-toxic, non-corrosive, non-channeling, generally recognized as safe (GRAS) by the FDA, not regulated by OSHA, economical to use, and can be regenerated repeatedly for reuse. Efficiency: The moisture remaining in gases dried with Drierite at 25° - 30° C is 0.005 mg./liter, (data verified by the National Bureau of Standards). Air is dried to a dew point of -100° F. Organic liquids are dried to the low ppm range. There is little change in efficiency at temperatures up to 100° C. Typical Analysis: >99.5% CaSO4