Learn More
Cole-Parmer Drierite Drying Tube, 300 mL/min, 4/pk
Supplier:  Cole-Parmer 0890300
Encompass_Preferred
Drierite Drying Tubes measure 8"L x " OD and come filled with 30 grams of indicating Drierite. Tubes dry to a dew point of 100degF with a flow rate up to 300 mL/min. Acrylic construction with polypropylene end caps. Inlet/outlet ports accept " to 3/8" ID tubing; 20 micron Porex filter is in each end.
Catalog No. NC9843596