Learn More
WA HAMMOND DRIERITE COMPANY DRIERITE, INDICATING 10-20 MESH 5 LB JAR, STOCK# 24005
Catalog No. NC3372169
Encompass_Preferred
Indicating Drierite is impregnated with cobalt chloride. Indicating Drierite is blue when dry and changes to pink upon absorption of moisture. The color change is pronounced and clearly visible. This makes Indicating Drierite valuable when it is necessary to know with certainty that dryness is being maintained and to signal when the drying agent should be replaced. It has the same efficiency as Regular Drierite, and somewhat greater capacity due to the desiccating effect of the cobalt chloride. It can be regenerated repeatedly for reuse. It should be noted that Indicating Drierite should not be used for the drying of ammonia, NH3, since ammonia reacts with inorganic chlorides. Efficiency: The moisture remaining in gases dried with Drierite at 25° - 30° C is 0.005 mg./liter, (data verified by the National Bureau of Standards). Air is dried to a dew point of -100° F. There is little change in efficiency at temperatures up to 100° C. Typical Analysis: ≥ 98% CaSO4, <2% CoCl2
Catalog No. NC3372169 Supplier WA HAMMOND DRIERITE COMPANY Supplier No. 24005
Indicating Drierite is impregnated with cobalt chloride. Indicating Drierite is blue when dry and changes to pink upon absorption of moisture. The color change is pronounced and clearly visible. This makes Indicating Drierite valuable when it is necessary to know with certainty that dryness is being maintained and to signal when the drying agent should be replaced. It has the same efficiency as Regular Drierite, and somewhat greater capacity due to the desiccating effect of the cobalt chloride. It can be regenerated repeatedly for reuse. It should be noted that Indicating Drierite should not be used for the drying of ammonia, NH3, since ammonia reacts with inorganic chlorides. Efficiency: The moisture remaining in gases dried with Drierite at 25° - 30° C is 0.005 mg./liter, (data verified by the National Bureau of Standards). Air is dried to a dew point of -100° F. There is little change in efficiency at temperatures up to 100° C. Typical Analysis: ≥ 98% CaSO4, <2% CoCl2