Learn More
Ace Glass, Inc. 8 MESH INDICATNG DRIERITE 5 LB
SDP
Catalog No. NC9306112
Encompass
8 mesh Drierite. The granules of calcium sulfate are impregnated with CP cobalt chloride. On dehydration, the active desiccant assumes a distinct blue color. In use, the color changes to a rose-red. 5/lb.
Catalog No. NC9306112 Supplier Ace Glass, Inc. Supplier No. 1017529
8 mesh Drierite. The granules of calcium sulfate are impregnated with CP cobalt chloride. On dehydration, the active desiccant assumes a distinct blue color. In use, the color changes to a rose-red. 5/lb.