Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIERITE™, Indicating, 4 Mesh, J.T. Baker™

Catalog No. 02002757 Shop All Avantor Products
Change view
Click to view available options
Quantity:
454 g
2.3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-002-757 2.3 kg
02-002-758 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-002-757 Supplier Avantor Supplier No. L05702
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Quantity 2.3 kg
Mesh Size 4 Mesh
Specific Gravity 1L = 1.87kg
Chemical Name or Material DRIERITE™, Indicating
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.