Learn More
Ace Glass, Inc. Drierite, indicating, 8 mesh
SDP
Catalog No. NC9979539
Encompass
The granules of calcium sulfate are impregnated with C.P. cobalt chloride. On dehydration, the active desiccant assumes a distinct blue color. In use, the color changes to a rose-red as the margin between the exhausted and active desiccant progresses through the tube or tower.
Catalog No. NC9979539 Supplier Ace Glass, Inc. Supplier No. 1017519
The granules of calcium sulfate are impregnated with C.P. cobalt chloride. On dehydration, the active desiccant assumes a distinct blue color. In use, the color changes to a rose-red as the margin between the exhausted and active desiccant progresses through the tube or tower.