Learn More
Andwin Scientific DRIERITE INDICATOR 4 MESH 1LB
SDP
Catalog No. NC9645956
Encompass_Preferred
The Indicating DRIERITE Absorbent is an effective drying agent that conveniently provides visual indication of the progress of absorption and exhaustion. This product can be utilized at the end of large units of regular DRIERITE to signify the activity or exhaustion of regular DRIERITE. This can absorb 6-10% of its weight in water, contingent on relative humidity. As this product absorbs moisture, the indicator's color will change from blue to rose-red. This has a mesh size of 4 and a weight of 0.45 kg (1 lb).
Catalog No. NC9645956 Supplier Andwin Scientific Supplier No. 22890900
The Indicating DRIERITE Absorbent is an effective drying agent that conveniently provides visual indication of the progress of absorption and exhaustion. This product can be utilized at the end of large units of regular DRIERITE to signify the activity or exhaustion of regular DRIERITE. This can absorb 6-10% of its weight in water, contingent on relative humidity. As this product absorbs moisture, the indicator's color will change from blue to rose-red. This has a mesh size of 4 and a weight of 0.45 kg (1 lb).