Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drierite™, Non-Indicating, Mesh Size: 8, MilliporeSigma™

Catalog No. mdx25101 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 lb.
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MDX25101 1 lb. Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MDX25101 Supplier MilliporeSigma Supplier No. DX25101
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
Synonym desiccant
PubChem CID 24497
ChEBI CHEBI:31346
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Color White
Melting Point 1450°C
Physical Form Irregular Granules
Quantity 1 lb.
Packaging Glass Bottle
Solubility Information 2g/L
Formula Weight 136.15
EINECSNumber 231-900-3
RTECSNumber WS6920000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.