Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drierite™, with indicator, 6 mesh

Catalog No. AC463475000
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463475000 500 g
AC463470020 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463475000 Supplier Thermo Scientific Chemicals Supplier No. 463475000

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
IUPAC Name calcium sulfate
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1450.0°C
Quantity 500 g
Solubility Information Solubility aq. soln.: slightly soluble. Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents
Formula Weight 136.13
Chemical Name or Material Drierite*
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May cause an allergic skin reaction.

May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Suspected of causing genetic defects.

May cause cancer by inhalation.

May damage fertility.<br/
Precautionary Statement GHS P Statement Obtain special instructions before use.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.

IF exposed or concerned: Get medical advice/attention.

EINECSNumber 231-900-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.