Learn More
Hach Company Drierite w/o indicator 454g
Supplier:  Hach Company 2285901
Encompass
Desiccant-Drierite-(without indicator) 454gIndicating Drierite changes from a blue color when dry to red upon absorption of moisture Packaged in 454-g (1 lb) containers
Catalog No. 50-003-04261