Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drierite™, without indicator, 8 mesh

Catalog No. AC219050020
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219050020 2 kg Plastic Bottle
AC219055000 500 g Glass Bottle
AC219050100 10 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC219050020 Supplier Thermo Scientific Chemicals Supplier No. 219050020
  • $220.00
    $175.65 / Each of 1

    Save $44.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1450.0°C
Physical Form Granular
Quantity 2 kg
Packaging Plastic Bottle
Solubility Information Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol
IUPAC Name calcium;sulfate
Formula Weight 136.14
Chemical Name or Material Drierite™

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.