Learn More
Sigma Aldrich Fine Chemicals Biosciences DrieriteTM gas-drying unit1EA
Catalog No. 502900482
Encompass
Molded-acrylic column (25/8 in. O.D. x 113/8 in.) filled with 11/4 pounds of 8-mesh indicating Drierite. Water-vapor capacity 50 g. Safe for working pressures up to 90 psig. flow rate should be 200 L/hr or 0.1 scfm for maximum efficiency. Regenerate by heating granules on a tray for 1 hr at 200 C. Hose barbs will fit 1/4 to 3/8 in. tubing.
Catalog No. 50-290-0482 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1128791EA
Molded-acrylic column (25/8 in. O.D. x 113/8 in.) filled with 11/4 pounds of 8-mesh indicating Drierite. Water-vapor capacity 50 g. Safe for working pressures up to 90 psig. flow rate should be 200 L/hr or 0.1 scfm for maximum efficiency. Regenerate by heating granules on a tray for 1 hr at 200 C. Hose barbs will fit 1/4 to 3/8 in. tubing.