Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ DH Air Vented Work Shirt
SDP

Catalog No. 17010912 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3X-Large
5X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-912 Medium
17-010-911 Large
17-010-913 X-Large
17-010-909 3X-Large
17-010-910 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-010-912 Supplier National Safety Apparel™ Supplier No. SHRA2TGVMDRG
Only null left
Add to Cart
Add to Cart

Work shirt features a patented vent design that actively allows hot air to exit the garment creating breathability. Covered back mesh panel wicks away sweat increasing mobility and performance.

  • Patented vent design with industry leading comfort
  • Lightweight and breathable fabric
  • Button down collar
  • Two front chest pockets with flaps and button closure
  • Quickly identify compliance with exterior labeling
  • Dual hazard protection against arc flash and flash fire

Specifications

Size Medium
Gender Unisex
Material Aramid, Lyocell, Modacrylic
Color Medium Blue
Neck Style Collar
Sleeve Style Long Sleeve
Closure Type Button
Disposable Reusable
Product Type Work Shirt
Hazard Risk Category CAT 2
Arc Flash Rating 9.1 cal/cm²
Certifications/Compliance NFPA 70E, NFPA 2112, ASTM F1506, CSA Z462, OSHA 1910.269
For Use With (Application) Utilities, Oil, Gas
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.