Learn More
National Safety Apparel™ DRIFIRE™ 4.4™ FR Three Layer Mask with Nose Wire
SDP
Catalog No. 17000525 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Standard
Catalog No. Size
17-000-525 Standard
1 options
Catalog No. 17-000-525 Supplier National Safety Apparel™ Supplier No. MASK2C44
Flame resistant 3-layer mask with nose wire is designed to discourage touching of the nose and mouth as well as to decrease the spread of airborne particles from coughing or sneezing.
  • Adjustable ear loops maximize comfort and optimize fit
  • Unsurpassed moisture wicking and dry rate
  • Lightweight, breathable fabric for maximum comfort
  • Inherently flame resistant; FR protection will not wash or wear out
  • Three Layer per CDC guidelines
  • Listed on CDC website as per NIOSH recommendations for Barrier Face Coverings
Specifications
Product Type Flame-resistant Face Mask
ASTM Level ASTM Level 1
Color Navy
Fastening Type Earloops
Material Aramid Fiber
Size Standard
Sterility Sterile
Style Pleated
Certifications/Compliance ASTM F3502, NFPA 70E, ASTM F1506
Disposable Reusable
Filter Efficiency 3-Layer per CDC guidelines
Latex Free Yes
Model MASK2C-44