Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ FR Control™ Cooling Neck Gaiter
SDP

Catalog No. 17010868 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-868 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-010-868 Supplier National Safety Apparel™ Supplier No. H01FK159
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to tackle the cold and heat, the FR Control™ Neck Gaiter is a versatile piece that regulates skin temperature, reduces perspiration. and offers heat stress reduction.

  • Temperature Regulation: Proactively adjusts skin temperature by +/- 3 degrees to keep you comfortable in both hot and cold environments
  • Stay Dry and Comfortable: Reduces perspiration by 34% with high-performance moisture-wicking and quick-drying fabric
  • Odor Control for Freshness: Odor-fighting technology keeps you feeling fresh throughout the day
  • Breathable Flame-Resistant Fabric: Made from lightweight, breathable FR material for added safety and comfort
  • Long-Lasting Protection: Inherently flame-resistant fabric that will not wash or wear out over time
  • Stretchable Fit: Flexible design stretches to move with you, ensuring a secure and comfortable fit

Specifications

Product Type FR Balaclava with Face Mask
Size One Size
Material Modacrylic, FR Rayon, Nylon, Spandex
Color Navy
Closure Type Pull-over
Disposable Reusable
Arc Flash Rating 4 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Includes Neck Gaiter
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Resistance Flame
Hood Type Unhooded
Style Neck Gaiter
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.