Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ FR Lightweight Long Sleeve T-Shirt
SDP

Catalog No. 17010865 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-865 Large
17-010-866 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-010-865 Supplier National Safety Apparel™ Supplier No. DF2CM446LSNBLG
Only null left
Add to Cart
Add to Cart

FR lightweight long sleeve t-shirt worn as a base layer or a stand-alone shirt offers lightweight comfort, odor control, and breathability.

  • Lightweight, breathable fabric
  • Moisture wicking and quick dry rate
  • Inherently flame resistant, FR protection will not wash/wear out
  • Odor control technology
  • Ribbed collar maintains shape
  • Ribbed cuff to hold sleeve in place
  • Soft fabric prevents chafing and maximizes comfort

Specifications

Size Large
Gender Unisex
Material Modacrylic Blend
Color Navy
Neck Style Crew
Sleeve Style Long Sleeve
Disposable Reusable
Product Type Long Sleeve Shirt
Hazard Risk Category CAT 1
Arc Flash Rating 4.5 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F1506, OSHA 1910.269, CSA Z462
Style Base Layer
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.