Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ FR Winter Beanie
SDP

Catalog No. 17010869 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-869 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-010-869 Supplier National Safety Apparel™ Supplier No. HMOD2BK
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Comfortable flame resistant headwear is designed for cold weather with a double layer knit to keep wearers warm.

  • Double layer knit
  • Stretchable fit for more comfort
  • Designed for cold weather conditions
  • Warmth without the weight
  • Inherently flame resistant, FR protection will not wash/wear out

Specifications

Product Type Fire Protectant Hood
Size One Size
Material FR Cotton, Nylon
Color Black
Closure Type Pull-over
Disposable Reusable
Arc Flash Rating 28 cal/cm²
Certifications/Compliance NFPA 70E. ASTM F1506, OSHA 1910.269, CSA Z462
Includes Winter Beanie
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Resistance Flame
Hood Type Unhooded
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.