Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ FR Winter Ear Band
SDP

Catalog No. 17010870 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-870 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-010-870 Supplier National Safety Apparel™ Supplier No. HNCEB
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Comfortable FR ear band is designed to fit over a hard hat with a double layer knit to keep wearers warm.

  • Item is worn over the hard hat to keep out wind
  • Double layer knit
  • Stretchable fit for more comfort
  • Designed for cold weather conditions
  • Warmth without the weight
  • Inherently flame resistant, FR protection will not wash/wear out

Specifications

Product Type Fire Protectant Hood
Size One Size
Material Nomex Knit Fabric
Color Black
Closure Type Pull-over
Disposable Reusable
Arc Flash Rating 35 cal/cm²
Certifications/Compliance NFPA 70E. ASTM F1506, OSHA 1910.269, CSA Z462
Includes Winter Ear Band
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Hood Type Unhooded
Style Ear Band
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.