Description
- Made from Polyoxymethylene (POM)
- Will not scratch glass
- Color-coded
- For standard taper joints
Specifications
Specifications
| Autoclavable | Autoclavable |
| Quantity | 10 Pk. |
| Type | Standard Taper Joint Clip |
| Material | POM |
| Size | 34 |
| Color | Orange |
| Jaw Type | Standard Taper |
| Sterility | Non-sterile |