Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ Power Dry ™ FR Long Sleeve Henley
SDP

Catalog No. 17010916 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Large
X-Large
3X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-916 3X-Large
17-010-914 Large
17-010-915 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-010-916 Supplier National Safety Apparel™ Supplier No. TEEPDGHN3X
Only null left
Add to Cart
Add to Cart

Polartec™ Power Dry™ FR henley knit shirt uses a patented tri-blend knit to deliver mechanical wicking action and high breathability for fast drying times and ultra-comfortable performance.

  • Dual Hazard protection against arc flash and flash fire
  • Lightweight and breathable fabric
  • Fast drying offers additional comfort
  • Left front chest pocket
  • Tagless neckline
  • Quickly identify compliance with exterior labeling

Specifications

Size 3X-Large
Gender Unisex
Material Aramid, Lyocell, Modacrylic
Color Gray
Neck Style Crew
Sleeve Style Long Sleeve
Disposable Reusable
Product Type Long Sleeve Shirt
Hazard Risk Category CAT 2
Arc Flash Rating 8.6 cal/cm²
Certifications/Compliance NFPA 70E, NFPA 2112, ASTM F1506, CSA Z462, OSHA 1910.269
Style Henley
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.