Learn More
Grainger Solo Drift Guard, Oval, Flat Spray Nozzle, 1 ea.
Catalog No. 502785200
Encompass_Preferred
This oval drift guard features an integrated flat spray nozzle designed to protect adjacent rows by minimizing spray drift and limiting the spray range. It is an essential tool for targeting weeds near cultivated plants and ponds.
  • Prevents drift of fine spray droplets
  • Limits spray application area
  • Supports a maximum operating pressure of 45 psi
  • Suitable for liquid temperatures up to 120°F
  • Compatible with specific manual sprayers including models 408, 409, 414, 453, 456, 457, 378, and 388
Catalog No. 50-278-5200 Supplier Grainger Supplier No. 190T39
This oval drift guard features an integrated flat spray nozzle designed to protect adjacent rows by minimizing spray drift and limiting the spray range. It is an essential tool for targeting weeds near cultivated plants and ponds.
  • Prevents drift of fine spray droplets
  • Limits spray application area
  • Supports a maximum operating pressure of 45 psi
  • Suitable for liquid temperatures up to 120°F
  • Compatible with specific manual sprayers including models 408, 409, 414, 453, 456, 457, 378, and 388