Learn More
MSC Genesis Ceiling Tile, Vinyl, 2 ft x 2 ft, White, Class A Fire Rated
Catalog No. 502837332
Encompass
This Genesis Drifts Vinyl Lay In Ceiling Tile features an undulating pattern and is made of durable, waterproof vinyl. Designed for easy installation into standard 15/16 inch grids, it provides a functional and aesthetic solution suitable for various commercial, healthcare, and retail environments.
  • Easy installation, just lay into any grid
  • Waterproof and washable
  • Mold and mildew resistant
  • Perfect for any damp or humid space
  • Sag resistant
  • Class A fire rating
  • Light reflectance of 74%
Catalog No. 50-283-7332 Supplier MSC Supplier No. 55663439
This Genesis Drifts Vinyl Lay In Ceiling Tile features an undulating pattern and is made of durable, waterproof vinyl. Designed for easy installation into standard 15/16 inch grids, it provides a functional and aesthetic solution suitable for various commercial, healthcare, and retail environments.
  • Easy installation, just lay into any grid
  • Waterproof and washable
  • Mold and mildew resistant
  • Perfect for any damp or humid space
  • Sag resistant
  • Class A fire rating
  • Light reflectance of 74%