Learn More
Grainger Drill and Tap Set 27/64 1/2 -13NC
Catalog No. 502752590
Encompass_Preferred
The WIDIA GTD Drill and Tap Set - 27/64" - 1/2"-13NC is a drill and tap set for creating 1/2"-13 threaded holes in metal. It includes a 27/64" high speed steel drill bit and a 1/2"-13NC high speed steel tap. The drill bit features a 135 degree split point for easy starting and clean holes. The tap has a spiral point for alignment and smooth cutting. Both are made from M2 high speed steel for durability. This set allows you to drill and tap 1/2"-13 threaded holes in one operation.
Catalog No. 50-275-2590 Supplier Grainger Supplier No. 141H71
The WIDIA GTD Drill and Tap Set - 27/64" - 1/2"-13NC is a drill and tap set for creating 1/2"-13 threaded holes in metal. It includes a 27/64" high speed steel drill bit and a 1/2"-13NC high speed steel tap. The drill bit features a 135 degree split point for easy starting and clean holes. The tap has a spiral point for alignment and smooth cutting. Both are made from M2 high speed steel for durability. This set allows you to drill and tap 1/2"-13 threaded holes in one operation.