Learn More
Grainger Drill and Tap Set 5/16 3/8-16NC
Catalog No. 502752589
Encompass_Preferred
The Grainger Approved Drill and Tap Set is a 29-piece set that includes high speed steel fractional taps and drills from #4-40 to 1/4"-20. It comes in a plastic index case for organization and portability. The taps have a taper length of 3-5 threads for hand tapping applications. The drills are 118 degree split point style for precise starting and clean holes. This set is ideal for general purpose tapping and drilling in metal, wood, and plastic. The index case keeps the pieces neatly organized for quick identification and access.
Catalog No. 50-275-2589 Supplier Grainger Supplier No. 141H70
The Grainger Approved Drill and Tap Set is a 29-piece set that includes high speed steel fractional taps and drills from #4-40 to 1/4"-20. It comes in a plastic index case for organization and portability. The taps have a taper length of 3-5 threads for hand tapping applications. The drills are 118 degree split point style for precise starting and clean holes. This set is ideal for general purpose tapping and drilling in metal, wood, and plastic. The index case keeps the pieces neatly organized for quick identification and access.