Learn More
Grainger Drill and Tap Set 6 -32NC Thread 2 pcs.
Catalog No. 502752382
Encompass_Preferred
The DEWALT Drill and Tap Set is a 15-piece set of high-speed steel fractional taps and drills. It includes plug, taper and bottoming taps from #4-40 to 1/4"-20 and the corresponding drill bits. The taps have a black oxide finish for corrosion resistance and the drill bits are made of high-speed steel for durability. This set is ideal for tapping holes in metal, plastic and wood. The compact plastic case keeps the set organized for storage and portability.
Catalog No. 50-275-2382 Supplier Grainger Supplier No. 141H63
The DEWALT Drill and Tap Set is a 15-piece set of high-speed steel fractional taps and drills. It includes plug, taper and bottoming taps from #4-40 to 1/4"-20 and the corresponding drill bits. The taps have a black oxide finish for corrosion resistance and the drill bits are made of high-speed steel for durability. This set is ideal for tapping holes in metal, plastic and wood. The compact plastic case keeps the set organized for storage and portability.